Assay Details
| Gene Symbol: UVR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125781.4 | NP_201191.1 | 2 - 3 | 577 | 78 | |
| GenBank mRNA | AY125497.1 | - | 179 | 78 | ||
| BT000600.1 | - | 1 - 2 | 96 | 78 | ||
More Information
Set Membership: |
|

UVR8
Genomic Map