Assay Details
| Gene Symbol: UVR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125781.4 | NP_201191.1 | 7 - 8 | 1318 | 66 | |
| GenBank mRNA | AY125497.1 | - | 920 | 66 | ||
| BT000600.1 | - | 6 - 7 | 837 | 66 | ||
More Information
Set Membership: |
|

UVR8
Genomic Map