Assay Details
| Gene Symbol: UVR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125781.4 | NP_201191.1 | 10 - 11 | 1588 | 63 | |
| GenBank mRNA | AY125497.1 | - | 1190 | 63 | ||
| BT000600.1 | - | 9 - 10 | 1107 | 63 | ||
More Information
Set Membership: |
|

UVR8
Genomic Map