Assay Details
| Gene Symbol: FCLY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125787.5 | NP_201196.1 | 4 - 5 | 1346 | 67 | |
| GenBank mRNA | AY081255.1 | - | 4 - 5 | 1255 | 67 | |
| AY128716.1 | - | 4 - 5 | 1245 | 67 | ||
More Information
Set Membership: |
|

FCLY
Genomic Map