Assay Details
| Gene Symbol: SAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125794.5 | NP_201203.3 | 3 - 4 | 221 | 122 | |
| GenBank mRNA | AK227460.1 | - | 3 - 4 | 337 | 122 | |
| AY034894.1 | - | 3 - 4 | 221 | 122 | ||
| BT005993.1 | - | 3 - 4 | 221 | 122 | ||
| U40433.1 | - | 3 - 4 | 328 | 122 | ||
More Information
Set Membership: |
|

SAL1
Genomic Map