Assay Details
| Gene Symbol: AT5G64170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085324.2 | NP_001078793.1 | 5 - 6 | 1848 | 74 | |
| NM_001345619.1 | NP_001330478.1 | 5 - 6 | 1848 | 74 | ||
| NM_125813.5 | NP_201222.1 | 4 - 5 | 1718 | 74 | ||
| GenBank mRNA | BT030360.1 | - | 3 - 4 | 1320 | 74 | |
More Information
Set Membership: |
|

AT5G64170
Genomic Map