Assay Details
| Gene Symbol: AT5G64220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037058.2 | NP_001032135.1 | 9 - 10 | 1667 | 99 | |
| NM_125818.3 | NP_201227.3 | 10 - 11 | 1478 | 99 | ||
| GenBank mRNA | AB493811.1 | - | 9 - 10 | 1426 | 99 | |
| AK229403.1 | - | 9 - 10 | 1652 | 99 | ||
| BT010874.1 | - | 9 - 10 | 1426 | 99 | ||
More Information
Set Membership: |
|

AT5G64220
Genomic Map