Assay Details
| Gene Symbol: AT5G64220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037058.2 | NP_001032135.1 | 7 - 8 | 906 | 111 | |
| NM_125818.3 | NP_201227.3 | 8 - 9 | 717 | 111 | ||
| GenBank mRNA | AB493811.1 | - | 7 - 8 | 665 | 111 | |
| AK229403.1 | - | 7 - 8 | 891 | 111 | ||
| BT010874.1 | - | 7 - 8 | 665 | 111 | ||
More Information
Set Membership: |
|

AT5G64220
Genomic Map