Assay Details
| Gene Symbol: BETA-UP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125833.5 | NP_201242.2 | 1 - 2 | 237 | 68 | |
| GenBank mRNA | AF465754.1 | - | 1 - 2 | 104 | 68 | |
| BT000451.1 | - | 1 - 2 | 104 | 68 | ||
| BT008503.1 | - | 1 - 2 | 104 | 68 | ||
More Information
Set Membership: |
|

BETA-UP
Genomic Map