Assay Details
| Gene Symbol: AT5G64490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125845.3 | NP_201254.1 | 1 | 952 | 139 | |
| GenBank mRNA | BT004172.1 | - | 1 | 952 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G64490| Gene Symbol: AT5G64490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125845.3 | NP_201254.1 | 1 | 952 | 139 | |
| GenBank mRNA | BT004172.1 | - | 1 | 952 | 139 | |
Set Membership: |
|