Assay Details
| Gene Symbol: CMPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125862.5 | NP_201271.1 | 1 | 1303 | 116 | |
| GenBank mRNA | BT023419.1 | - | 1 | 1200 | 116 | |
| BT025629.1 | - | 1 | 1129 | 116 | ||
More Information
Set Membership: |
|

CMPG2
Genomic Map