Assay Details
| Gene Symbol: CESA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125870.3 | NP_201279.1 | 1 - 2 | 600 | 90 | |
| GenBank mRNA | KJ676983.1 | - | 1 - 2 | 78 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CESA6| Gene Symbol: CESA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125870.3 | NP_201279.1 | 1 - 2 | 600 | 90 | |
| GenBank mRNA | KJ676983.1 | - | 1 - 2 | 78 | 90 | |
Set Membership: |
|