Assay Details
| Gene Symbol: CESA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125870.3 | NP_201279.1 | 7 - 8 | 2034 | 68 | |
| GenBank mRNA | KJ676983.1 | - | 7 - 8 | 1512 | 68 | |
More Information
Set Membership: |
|

CESA6
Genomic Map