Assay Details
| Gene Symbol: LIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345650.1 | NP_001318878.1 | 2 - 3 | 807 | 91 | |
| NM_001345651.1 | NP_001331706.1 | 3 - 4 | 723 | 91 | ||
| NM_125878.3 | NP_568996.1 | 1 - 2 | 792 | 91 | ||
| GenBank mRNA | AF360203.1 | - | 1 - 2 | 589 | 91 | |
| AY040050.1 | - | 1 - 2 | 94 | 91 | ||
More Information
Set Membership: |
|

LIP1
Genomic Map