Assay Details
| Gene Symbol: LIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345650.1 | NP_001318878.1 | 6 - 7 | 1403 | 93 | |
| NM_001345651.1 | NP_001331706.1 | 7 - 8 | 1319 | 93 | ||
| NM_125878.3 | NP_568996.1 | 5 - 6 | 1388 | 93 | ||
| GenBank mRNA | AF360203.1 | - | 5 - 6 | 1185 | 93 | |
| AY040050.1 | - | 5 - 6 | 690 | 93 | ||
More Information
Set Membership: |
|

LIP1
Genomic Map