Assay Details
| Gene Symbol: ABCF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125882.3 | NP_201289.1 | 4 - 5 | 2052 | 92 | |
| GenBank mRNA | AK227200.1 | - | 4 - 5 | 1574 | 92 | |
More Information
Set Membership: |
|

ABCF5
Genomic Map