Assay Details
| Gene Symbol: CPR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125892.4 | NP_569003.1 | 1 - 2 | 461 | 61 | |
| GenBank mRNA | AY033229.1 | - | 1 - 2 | 385 | 61 | |
| BT033151.1 | - | 1 - 2 | 385 | 61 | ||
More Information
Set Membership: |
|

CPR5
Genomic Map