Assay Details
| Gene Symbol: ANNAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161326.2 | NP_001154798.1 | 3 - 4 | 1005 | 68 | |
| NM_125901.4 | NP_201307.1 | 4 - 5 | 763 | 68 | ||
| GenBank mRNA | AF083914.1 | - | 4 - 5 | 709 | 68 | |
| AY070400.1 | - | 4 - 5 | 746 | 68 | ||
| AY085713.1 | - | 4 - 5 | 747 | 68 | ||
| AY096577.1 | - | 4 - 5 | 652 | 68 | ||
More Information
Set Membership: |
|

ANNAT2
Genomic Map