Assay Details
| Gene Symbol: MAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203688.1 | NP_001190617.1 | 3 - 4 | 405 | 100 | |
| NM_001345664.1 | NP_001332255.1 | 2 - 3 | 358 | 100 | ||
| NM_001345666.1 | NP_001332257.1 | 1 - 2 | 196 | 100 | ||
| NM_125906.4 | NP_201312.2 | 2 - 3 | 359 | 100 | ||
| GenBank mRNA | AY231450.1 | - | 2 - 3 | 292 | 100 | |
More Information
Set Membership: |
|

MAF4
Genomic Map