Assay Details
| Gene Symbol: MAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126027.1 | NP_001119499.1 | 4 - 5 | 441 | 98 | |
| NM_001203688.1 | NP_001190617.1 | 4 - 5 | 461 | 98 | ||
| NM_001345664.1 | NP_001332255.1 | 3 - 4 | 414 | 98 | ||
| NM_001345665.1 | NP_001332256.1 | 4 - 5 | 441 | 98 | ||
| NM_001345666.1 | NP_001332257.1 | 2 - 3 | 252 | 98 | ||
| NM_125906.4 | NP_201312.2 | 3 - 4 | 415 | 98 | ||
| GenBank mRNA | AY231450.1 | - | 3 - 4 | 348 | 98 | |
More Information
Set Membership: |
|

MAF4
Genomic Map