Assay Details
| Gene Symbol: BST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126028.2 | NP_001119500.1 | 2 - 3 | 880 | 65 | |
| NM_001345669.1 | NP_001318880.1 | 6 - 7 | 1085 | 65 | ||
| NM_125908.5 | NP_201314.3 | 5 - 6 | 1078 | 65 | ||
| GenBank mRNA | BT015346.1 | - | 4 - 5 | 977 | 65 | |
| BT015797.1 | - | 4 - 5 | 915 | 65 | ||
More Information
Set Membership: |
|

BST1
Genomic Map