Assay Details
| Gene Symbol: AT5G65620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345702.1 | NP_001332654.1 | 15 - 16 | 2341 | 82 | |
| NM_125960.3 | NP_569013.1 | 15 - 16 | 2238 | 82 | ||
| GenBank mRNA | AY045936.1 | - | 15 - 16 | 2222 | 82 | |
| AY142682.1 | - | 15 - 16 | 2219 | 82 | ||
More Information
Set Membership: |
|

AT5G65620
Genomic Map