Assay Details
| Gene Symbol: ATBCAT-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125975.8 | NP_201379.2 | 9 - 10 | 1780 | 85 | |
| GenBank mRNA | AJ293804.1 | - | 7 - 8 | 1492 | 85 | |
| AK228603.1 | - | 7 - 8 | 951 | 85 | ||
More Information
Set Membership: |
|

ATBCAT-5
Genomic Map