Assay Details
| Gene Symbol: ACS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125977.2 | NP_201381.1 | 1 - 2 | 398 | 71 | |
| GenBank mRNA | AF334720.1 | - | 1 - 2 | 284 | 71 | |
| AK229087.1 | - | 1 - 2 | 398 | 71 | ||
| L29261.1 | - | 1 - 2 | 284 | 71 | ||
More Information
Set Membership: |
|

ACS5
Genomic Map