Assay Details
| Gene Symbol: AT5G65900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125987.3 | NP_201391.1 | 5 - 6 | 1208 | 100 | |
| GenBank mRNA | AY059930.1 | - | 5 - 6 | 1165 | 100 | |
| BT001188.1 | - | 5 - 6 | 1041 | 100 | ||
More Information
Set Membership: |
|

AT5G65900
Genomic Map