Assay Details
| Gene Symbol: AT1G43666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202239.2 | NP_973968.1 | 1 | 458 | 114 | |
| GenBank mRNA | AK221433.1 | - | 1 | 458 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G43666| Gene Symbol: AT1G43666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202239.2 | NP_973968.1 | 1 | 458 | 114 | |
| GenBank mRNA | AK221433.1 | - | 1 | 458 | 114 | |
Set Membership: |
|