Assay Details
| Gene Symbol: BGLU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333247.1 | NP_001323171.1 | 4 - 5 | 621 | 151 | |
| NM_001333248.1 | NP_001323172.1 | 4 - 5 | 572 | 151 | ||
| NM_001333249.1 | NP_001323173.1 | 4 - 5 | 339 | 151 | ||
| NM_001333250.1 | NP_001323174.1 | 4 - 5 | 339 | 151 | ||
| NM_179440.2 | NP_849771.2 | 5 - 6 | 375 | 151 | ||
| NM_202245.2 | NP_973974.2 | 4 - 5 | 339 | 151 | ||
More Information
Set Membership: |
|

BGLU1
Genomic Map