Assay Details
| Gene Symbol: MLO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123796.1 | NP_001117268.1 | 7 - 8 | 1194 | 113 | |
| NM_001331977.1 | NP_001320358.1 | 6 - 7 | 892 | 113 | ||
| NM_101004.4 | NP_172598.1 | 7 - 8 | 1242 | 113 | ||
| GenBank mRNA | AF369563.1 | - | 7 - 8 | 942 | 113 | |
| AK226449.1 | - | 7 - 8 | 1195 | 113 | ||
| AY086586.1 | - | 7 - 8 | 1153 | 113 | ||
More Information
Set Membership: |
|

MLO2
Genomic Map