Assay Details
| Gene Symbol: AT1G11380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101011.3 | NP_563889.1 | 1 - 2 | 448 | 101 | |
| GenBank mRNA | AY084625.1 | - | 1 - 2 | 388 | 101 | |
| BT024901.1 | - | 1 - 2 | 316 | 101 | ||
More Information
Set Membership: |
|

AT1G11380
Genomic Map