Assay Details
| Gene Symbol: CYP51G1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101040.4 | NP_172633.1 | 2 - 3 | 743 | 103 | |
| GenBank mRNA | AB014459.1 | - | 2 - 3 | 678 | 103 | |
| AF410338.1 | - | 2 - 3 | 645 | 103 | ||
| AX765850.1 | - | 1 - 2 | 468 | 103 | ||
| AY050860.1 | - | 2 - 3 | 645 | 103 | ||
| AY084517.1 | - | 2 - 3 | 585 | 103 | ||
| AY091203.1 | - | 1 - 2 | 468 | 103 | ||
More Information
Set Membership: |
|

CYP51G1
Genomic Map