Assay Details
| Gene Symbol: SS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198036.2 | NP_001184965.1 | 13 - 14 | 3151 | 86 | |
| NM_101044.3 | NP_172637.2 | 12 - 13 | 3040 | 86 | ||
More Information
Set Membership: |
|

SS3
Genomic Map