Assay Details
| Gene Symbol: SS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198036.2 | NP_001184965.1 | 15 - 16 | 3381 | 71 | |
| NM_101044.3 | NP_172637.2 | 14 - 15 | 3270 | 71 | ||
More Information
Set Membership: |
|

SS3
Genomic Map