Assay Details
| Gene Symbol: SS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198036.2 | NP_001184965.1 | 4 - 5 | 1753 | 118 | |
| NM_101044.3 | NP_172637.2 | 3 - 4 | 1642 | 118 | ||
More Information
Set Membership: |
|

SS3
Genomic Map