Assay Details
| Gene Symbol: CLPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101047.3 | NP_563893.1 | 3 - 4 | 339 | 133 | |
| GenBank mRNA | AF348580.1 | - | 3 - 4 | 206 | 133 | |
| AK229049.1 | - | 3 - 4 | 268 | 133 | ||
| AY087248.1 | - | 3 - 4 | 337 | 133 | ||
More Information
Set Membership: |
|

CLPP6
Genomic Map