Assay Details
| Gene Symbol: CLPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198037.1 | NP_001184966.1 | 6 - 7 | 655 | 74 | |
| NM_101047.3 | NP_563893.1 | 5 - 6 | 608 | 74 | ||
| GenBank mRNA | AF348580.1 | - | 5 - 6 | 475 | 74 | |
| AK229049.1 | - | 5 - 6 | 537 | 74 | ||
| AY087248.1 | - | 5 - 6 | 606 | 74 | ||
More Information
Set Membership: |
|

CLPP6
Genomic Map