Assay Details
| Gene Symbol: CLPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198037.1 | NP_001184966.1 | 8 - 9 | 852 | 72 | |
| NM_101047.3 | NP_563893.1 | 7 - 8 | 805 | 72 | ||
| GenBank mRNA | AF348580.1 | - | 7 - 8 | 672 | 72 | |
| AK229049.1 | - | 7 - 8 | 734 | 72 | ||
| AY087248.1 | - | 7 - 8 | 803 | 72 | ||
More Information
Set Membership: |
|

CLPP6
Genomic Map