Assay Details
| Gene Symbol: AT1G11780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101050.4 | NP_172643.1 | 5 - 6 | 831 | 85 | |
| GenBank mRNA | BT030097.1 | - | 5 - 6 | 771 | 85 | |
More Information
Set Membership: |
|

AT1G11780
Genomic Map