Assay Details
| Gene Symbol: ADT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035947.1 | NP_001031024.1 | 1 - 2 | 495 | 99 | |
| NM_101051.3 | NP_172644.1 | 1 - 2 | 590 | 99 | ||
| GenBank mRNA | DQ411466.1 | - | 1 - 2 | 271 | 99 | |
More Information
Set Membership: |
|

ADT1
Genomic Map