Assay Details
| Gene Symbol: ADT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035947.1 | NP_001031024.1 | 2 - 3 | 554 | 84 | |
| NM_101051.3 | NP_172644.1 | 2 - 3 | 649 | 84 | ||
| GenBank mRNA | DQ411466.1 | - | 2 - 3 | 330 | 84 | |
More Information
Set Membership: |
|

ADT1
Genomic Map