Assay Details
| Gene Symbol: AT1G12010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101073.3 | NP_172665.1 | 2 - 3 | 451 | 92 | |
| GenBank mRNA | AF446874.1 | - | 2 - 3 | 342 | 92 | |
| AY052694.1 | - | 2 - 3 | 409 | 92 | ||
More Information
Set Membership: |
|

AT1G12010
Genomic Map