Assay Details
| Gene Symbol: SUMM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332027.1 | NP_001322571.1 | 3 | 2534 | 72 | |
| NM_101100.2 | NP_172692.1 | 1 | 2750 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUMM2| Gene Symbol: SUMM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332027.1 | NP_001322571.1 | 3 | 2534 | 72 | |
| NM_101100.2 | NP_172692.1 | 1 | 2750 | 72 | ||
Set Membership: |
|