Assay Details
| Gene Symbol: ACR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101114.4 | NP_172704.1 | 1 - 2 | 358 | 94 | |
| GenBank mRNA | AF528064.1 | - | 1 - 2 | 102 | 94 | |
| AK228687.1 | - | 1 - 2 | 275 | 94 | ||
| BT026518.1 | - | 1 - 2 | 64 | 94 | ||
More Information
Set Membership: |
|

ACR8
Genomic Map