Assay Details
| Gene Symbol: ARK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101115.3 | NP_563908.1 | 16 - 17 | 2705 | 82 | |
| GenBank mRNA | AB290930.1 | - | 16 - 17 | 2479 | 82 | |
| AF159052.1 | - | 16 - 17 | 2669 | 82 | ||
More Information
Set Membership: |
|

ARK3
Genomic Map