Assay Details
| Gene Symbol: ARK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198043.1 | NP_001184972.1 | 2 - 3 | 517 | 107 | |
| NM_101115.3 | NP_563908.1 | 2 - 3 | 548 | 107 | ||
| GenBank mRNA | AB290930.1 | - | 2 - 3 | 322 | 107 | |
| AF159052.1 | - | 2 - 3 | 512 | 107 | ||
More Information
Set Membership: |
|

ARK3
Genomic Map