Assay Details
| Gene Symbol: EXPA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101127.4 | NP_172717.1 | 2 - 3 | 534 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EXPA7| Gene Symbol: EXPA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101127.4 | NP_172717.1 | 2 - 3 | 534 | 113 | |
Set Membership: |
|