Assay Details
| Gene Symbol: RBL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198046.2 | NP_001184975.1 | 5 - 6 | 1026 | 68 | |
| NM_001332047.1 | NP_001318990.1 | 4 - 5 | 1452 | 68 | ||
| NM_001332048.1 | NP_001322586.1 | 4 - 5 | 1476 | 68 | ||
| NM_001332049.1 | NP_001322585.1 | 4 - 5 | 1452 | 68 | ||
| NM_001332050.1 | NP_001322584.1 | 4 - 5 | 854 | 68 | ||
| NM_001332051.1 | NP_001322583.1 | 3 - 4 | 536 | 68 | ||
| NM_101145.3 | NP_172735.1 | 5 - 6 | 773 | 68 | ||
| GenBank mRNA | AY065251.1 | - | 5 - 6 | 710 | 68 | |
| AY091318.1 | - | 2 - 3 | 395 | 68 | ||
More Information
Set Membership: |
|

RBL6
Genomic Map