Assay Details
| Gene Symbol: UGE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101148.4 | NP_172738.1 | 5 - 6 | 1573 | 68 | |
| GenBank mRNA | AF334724.1 | - | 5 - 6 | 799 | 68 | |
| AY054207.1 | - | 5 - 6 | 903 | 68 | ||
| AY120709.1 | - | 5 - 6 | 1009 | 68 | ||
| BT000032.1 | - | 5 - 6 | 799 | 68 | ||
More Information
Set Membership: |
|

UGE1
Genomic Map