Assay Details
| Gene Symbol: DET3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101154.4 | NP_563916.1 | 2 - 3 | 1566 | 62 | |
| GenBank mRNA | AB493453.1 | - | 2 - 3 | 187 | 62 | |
| AF208261.1 | - | 2 - 3 | 196 | 62 | ||
| AF334725.1 | - | 2 - 3 | 187 | 62 | ||
| AY059872.1 | - | 2 - 3 | 254 | 62 | ||
| AY093334.1 | - | 2 - 3 | 187 | 62 | ||
More Information
Set Membership: |
|

DET3
Genomic Map