Assay Details
| Gene Symbol: DET3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101154.4 | NP_563916.1 | 7 - 8 | 2120 | 69 | |
| GenBank mRNA | AB493453.1 | - | 7 - 8 | 741 | 69 | |
| AF208261.1 | - | 7 - 8 | 750 | 69 | ||
| AF334725.1 | - | 7 - 8 | 741 | 69 | ||
| AY059872.1 | - | 7 - 8 | 808 | 69 | ||
| AY093334.1 | - | 7 - 8 | 741 | 69 | ||
More Information
Set Membership: |
|

DET3
Genomic Map