Assay Details
| Gene Symbol: SCRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332053.1 | NP_001320453.1 | 2 - 3 | 1550 | 77 | |
| NM_001332054.1 | NP_001320452.1 | 1 - 2 | 1617 | 77 | ||
| NM_101157.3 | NP_172746.2 | 2 - 3 | 1550 | 77 | ||
| GenBank mRNA | AK118011.1 | - | 2 - 3 | 1167 | 77 | |
| BT005377.1 | - | 2 - 3 | 1096 | 77 | ||
| EU369670.1 | - | 2 - 3 | 1231 | 77 | ||
More Information
Set Membership: |
|

SCRM2
Genomic Map